Skip to Content
Merck
(±)-exo,exo-2,3-Camphanediol 97%

(±)-exo,exo-2,3-Camphanediol

(±)-exo,exo-2,3-Camphanediol 97%
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing