Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-2-Aminobutyric acid
Linear Formula:
C
2
H
5
CH(NH
2
)CO
2
H
CAS Number:
2835-81-6
Molecular Weight:
103.12
EC Number:
220-616-5
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
162663
DL-2-Aminobutyric acid,
99%, for peptide synthesis,
ReagentPlus
®
Expand
View Pricing
67338
DL-2-Aminobutyric acid,
analytical standard
Expand
View Pricing