Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diisopropylamine
Synonym(s):
Diisopropylamine, DIPA
Linear Formula:
(CH
3
)
2
CHNHCH(CH
3
)
2
CAS Number:
108-18-9
Molecular Weight:
101.19
EC Number:
203-558-5
Products
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
8.03646
Diisopropylamine,
for synthesis
Expand
View Pricing
471224
Diisopropylamine,
≥99.5%
Expand
View Pricing
386464
Diisopropylamine,
purified by redistillation, 99.95%
Expand
View Pricing
38290
Diisopropylamine,
puriss. p.a., ≥99.0% (GC)
Expand
View Pricing
38292
Diisopropylamine,
analytical standard
Expand
View Pricing