Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diethylene glycol monoethyl ether acetate
Synonym(s):
2-(2-Ethoxyethoxy)ethyl acetate, Ethyldiglycol acetate
Linear Formula:
C
2
H
5
OCH
2
CH
2
OCH
2
CH
2
CO
2
CH
3
CAS Number:
112-15-2
Molecular Weight:
176.21
EC Number:
203-940-1
Products
Product Category
building blocks (1)
solvents (1)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
537527
Diethylene glycol monoethyl ether acetate,
99%
Expand
View Pricing
32240
Diethylene glycol monoethyl ether acetate,
≥99.0% (GC)
Expand
View Pricing