
Diclofenac Impurity C

Synonym(s):
{2-[(2,6-Dichlorophenyl)amino]phenyl}methanol, 2-[(2,6-Dichlorophenyl)amino]benzenemethanol, o-(2,6-Dichloroanilino)benzyl alcohol
Empirical Formula (Hill Notation):
C13H11Cl2NO
CAS Number:
Molecular Weight:
268.14
Products
Brand
Formula Weight
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing