Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dichloromethane-d
2
Synonym(s):
Dideuteromethylenechloride, Methylene chloride-d
2
, Dichloromethane-D2
Linear Formula:
CD
2
Cl
2
CAS Number:
1665-00-5
Molecular Weight:
86.94
EC Number:
216-776-0
Products
Product Category
solvents (5)
stable isotopes (2)
Brand
Sigma-Aldrich (6)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Isotope
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
444324
Dichloromethane-d
2
,
99.9 atom % D
Expand
View Pricing
177865
Dichloromethane-d
2
,
99.5 atom % D
Expand
View Pricing
233366
Dichloromethane-d
2
,
"100%", 99.96 atom % D
Expand
View Pricing
530506
Dichloromethane-d
2
,
99.9 atom % D, contains 0.1 % (v/v) TMS
Expand
View Pricing
1.13720
Dichloromethane-d
2
,
deuteration degree min. 99.8% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
296163
Dichloromethane-d
2
,
≥99.5 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing