
D-Glucose-d12

Synonym(s):
Dextrose-d12
Empirical Formula (Hill Notation):
C6D12O6
Molecular Weight:
192.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing