Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Deep well plate
Products
Product Category
plates (12)
Color
Manufacturer
Binding Type
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
AXYPDW10ML24CS
Deep well plate,
with rectangular well, clear, size 24 wells, size 10 mL, pkg of 5x5plates/cs, sterile
Expand
View Pricing
AXYP2MLSQC
Deep well plate,
96 square wells, working volume 2.0 mL, 25 ea of
Expand
View Pricing
AXYPDW20CS
Deep well plate,
size 96 wells, size 2.0 mL, pkg of 10x5plates/cs, sterile
Expand
View Pricing
AXYP2MLSQCS
Deep well plate,
25 ea of, sterile
Expand
View Pricing
AXYPDW10ML24C
Deep well plate,
with rectangular well, size 24 wells, clear, size 10 mL, pkg of 5x5plates/cs, non-sterile
Expand
View Pricing
AXYPDW11CS
Deep well plate,
size 96 wells, size 1.1 mL, pkg of 10x5plates/cs, sterile
Expand
View Pricing
AXYPDW11C
Deep well plate,
size 96 wells, size 1.1 mL, pkg of 10x5plates/cs
Expand
View Pricing
AXYPDW20C
Deep well plate,
size 96 wells, size 2.0 mL, pkg of 10x5plates/cs
Expand
View Pricing
AXYPDW20CSIND
Deep well plate,
individually wrapped, size 96 wells, size 2.0 mL, pkg of 30 ea, sterile
Expand
View Pricing
AXYPDW500CS
Deep well plate,
for SBS FootPrint, size 96 wells, size 600 μL, pkg of 10x5plates/cs, sterile
Expand
View Pricing
AXYPDW500CSI
Deep well plate,
for SBS FootPrint, individually wrapped, size 96 wells, size 600 μL, 25 ea of, sterile
Expand
View Pricing
AXYPDW500C
Deep well plate,
for SBS FootPrint, size 96 wells, size 600 μL, pkg of 10x5plates/cs
Expand
View Pricing