Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-(+)-Mannose
Synonym(s):
D-Mannopyranose
Empirical Formula (Hill Notation):
C
6
H
12
O
6
CAS Number:
3458-28-4
Molecular Weight:
180.16
EC Number:
222-392-4
Products
Product Category
carbohydrates (7)
monosaccharides (2)
Brand
Millipore (2)
Sigma-Aldrich (3)
Supelco (1)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
M6020
D-(+)-Mannose,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
M8574
D-(+)-Mannose,
synthetic, ≥99% (GC)
Expand
View Pricing
M2069
D-(+)-Mannose,
≥99% (GC), wood
Expand
View Pricing
63579
D-(+)-Mannose,
BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Expand
View Pricing
63580
D-(+)-Mannose,
suitable for microbiology, ≥99%
Expand
View Pricing
92683
D-(+)-Mannose,
analytical standard
Expand
View Pricing
63582
D-(+)-Mannose,
≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Expand
View Pricing