Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Crimp seals with PTFE/silicone septa
Products
Product Category
septa (12)
Brand
Supelco (12)
Diameter (mm)
OD (mm)
Closure Type
Color
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
27362
Crimp seals with PTFE/silicone septa,
silver aluminum seal, seal diam. 20 mm, open center, white PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
854996
Crimp seals with PTFE/silicone septa,
silver aluminum seal, open center (10 mm opening), diam. × thickness 20 mm × 3.25 mm, tan PTFE/silicone, pkg of 100 ea
Expand
View Pricing
27229
Crimp seals with PTFE/silicone septa,
silver aluminum seal, open center, diam. × total thickness × PTFE thickness 20 mm × 3.2 mm × 2 mil, white PTFE/silicone, pkg of 100 ea
Expand
View Pricing
27455-U
Crimp seals with PTFE/silicone septa,
silver aluminum seal,
pressure release open center
, PTFE/silicone, diam. × total thickness × PTFE thickness 20 mm × 2.5 mm × 5 mil, pkg of 100 ea
Expand
View Pricing
SU860010
Crimp seals with PTFE/silicone septa,
diam. × total thickness × PTFE thickness 20 mm × 3 mm × 5 mil, silver aluminum seal (10mm opening), open center, (silicone blue transparent/PTFE transparent), pkg of 1000 ea
Expand
View Pricing
27214-U
Crimp seals with PTFE/silicone septa,
silver aluminum seal, size × thickness 11 mm × 1.0 mm, white PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
27373
Crimp seals with PTFE/silicone septa,
silver aluminum seal, seal diam. 11 mm, red PTFE/silicone septum, pkg of 1000 ea
Expand
View Pricing
27360-U
Crimp seals with PTFE/silicone septa,
silver aluminum seal, diam. × thickness 11 mm × 1.0 mm, red PTFE/silicone septum, pkg of × 100 ea
Expand
View Pricing
508489
Crimp seals with PTFE/silicone septa,
aluminum seal (open center), diam. × thickness 20 mm × 2.9 mm, PTFE/silicone (blue silicone), Chromacol 20-AC-ST3, pkg of 100 ea
Expand
View Pricing
27375
Crimp seals with PTFE/silicone septa,
silver aluminum seal, seal diam. 20 mm, open center, white PTFE/silicone septum, pkg of 1000 ea
Expand
View Pricing
27452
Crimp seals with PTFE/silicone septa,
silver aluminum seal (open center), seal diam. 13 mm, PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
27215
Crimp seals with PTFE/silicone septa,
silver aluminum seal, seal diam. 11 mm, white PTFE/silicone septum
Expand
View Pricing