Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corticosterone
Synonym(s):
11β,21-Dihydroxy-4-pregnene-3,20-dione, 11β,21-Dihydroxyprogesterone, 4-Pregnene-11β,21-diol-3,20-dione, Kendall’s Compound B, Reichstein’s Substance H, Corticosterone
Empirical Formula (Hill Notation):
C
21
H
30
O
4
CAS Number:
50-22-6
Molecular Weight:
346.46
EC Number:
200-019-6
Products
Product Category
bioactive small molecules (2)
hormones (2)
lipids (1)
metabolites (1)
steroids (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Puriss (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
27840
Corticosterone,
≥98.5% (HPLC)
Expand
View Pricing
C2505
Corticosterone,
≥92% (HPLC), powder, corticosteroid hormone
Expand
View Pricing
235135
Corticosterone,
≥95% (HPLC), crystalline powder, corticosteroid hormone, Calbiochem
Expand
View Pricing
46148
Corticosterone,
VETRANAL
®
, analytical standard
Expand
View Pricing