Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Thermowell GOLD PCR 96 well plates
Products
Product Category
plates (4)
Color
Manufacturer
Volume (µL)
Wells
Application
Color
Plate Material
Package Size
Sterility
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS3752
Corning
®
Thermowell GOLD PCR 96 well plates,
96 well plate, Thermowell PCR GOLD plate, full skirt, polypropylene, conical bottom, clear, non-sterile, 50/cs
Expand
View Pricing
CLS3753
Corning
®
Thermowell GOLD PCR 96 well plates,
96 well plate, Thermowell GOLD PCR plate, half skirt, polypropylene, conical bottom, clear, non-sterile, 50/cs
Expand
View Pricing
CLS3754
Corning
®
Thermowell GOLD PCR 96 well plates,
96 well plate, Thermowell PCR GOLD plate, elevated skirt, polypropylene, conical bottom, clear, non-sterile, 50/cs
Expand
View Pricing
CLS3758
Corning
®
Thermowell GOLD PCR 96 well plates,
96 well plate, Thermowell PCR GOLD plate, half skirt, polypropylene, conical bottom, clear, non-sterile, w/ 2 generic barcode labels, 50/cs
Expand
View Pricing