Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Isotip
®
filtered pipet tips
Products
Product Category
pipette tips (7)
Manufacturer
Volume Range (µL)
Color
Package Size
Special Grade
Natural (7)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS4809
Corning
®
Isotip
®
filtered pipet tips,
100-1000 μL, universal fit racked pipet tips, natural, sterile, 100 tips/rack, 1000 tips/case
Expand
View Pricing
CLS4823
Corning
®
Isotip
®
filtered pipet tips,
1-200 uL, 2 inches long, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Expand
View Pricing
CLS4807
Corning
®
Isotip
®
filtered pipet tips,
0.2-10 μL, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Expand
View Pricing
CLS4810
Corning
®
Isotip
®
filtered pipet tips,
1-200 μL, 3 inches long, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Expand
View Pricing
CLS4821
Corning
®
Isotip
®
filtered pipet tips,
1-30 μL, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Expand
View Pricing
CLS4801
Corning
®
Isotip
®
filtered pipet tips,
0.1-2.0 μL, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Expand
View Pricing
CLS4808
Corning
®
Isotip
®
filtered pipet tips,
0.5-10 μL, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Expand
View Pricing