Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Ice Pan, rectangular
Products
Color
Manufacturer
Length (cm)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
CLS432094
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, blue
Expand
View Pricing
CLS432104
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, blue
Expand
View Pricing
CLS432107
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, lime green
Expand
View Pricing
CLS432098
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, pink
Expand
View Pricing
CLS432108
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, pink
Expand
View Pricing
CLS432105
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, red
Expand
View Pricing
CLS432103
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, green
Expand
View Pricing
CLS432109
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, purple
Expand
View Pricing
CLS432106
Corning
®
Ice Pan, rectangular,
midi, capacity 4 L, orange
Expand
View Pricing
CLS432093
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, green
Expand
View Pricing
CLS432099
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, purple
Expand
View Pricing
CLS432097
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, lime green
Expand
View Pricing
CLS432096
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, orange
Expand
View Pricing
CLS432095
Corning
®
Ice Pan, rectangular,
maxi, capacity 9 L, red
Expand
View Pricing