Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Falcon
®
Cell Strainer For Use With 50ml Conical Tubes
Products
Manufacturer
Application
Color
Pore size (µm)
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS352350
Corning
®
Falcon
®
Cell Strainer For Use With 50ml Conical Tubes,
pore size 70 μm, white Nylon, sterile
Expand
View Pricing
CLS352340
Corning
®
Falcon
®
Cell Strainer For Use With 50ml Conical Tubes,
pore size 40 μm, blue Nylon, sterile
Expand
View Pricing
CLS352360
Corning
®
Falcon
®
Cell Strainer For Use With 50ml Conical Tubes,
pore size 100 μm, yellow Nylon, sterile
Expand
View Pricing
CLS358339
Corning
®
Falcon
®
Cell Strainer For Use With 50ml Conical Tubes,
size 40 μm, blue Nylon, non-sterile
Expand
View Pricing
CLS358037
Corning
®
Falcon
®
Cell Strainer For Use With 50ml Conical Tubes,
size 40 μm, double bagged, non-sterile
Expand
View Pricing