Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Erlenmeyer cell culture flasks
Products
Product Category
flasks (12)
Manufacturer
Capacity (L)
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Sterilization Method
Style
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
CLS431143
Corning
®
Erlenmeyer cell culture flasks,
125 mL Erlenmeyer Flask w/ Vent Cap, polycarbonate, sterile
Expand
View Pricing
CLS431147
Corning
®
Erlenmeyer cell culture flasks,
1 L Erlenmeyer Flask w/ Vent Cap, polycarbonate, sterile, 25/cs
Expand
View Pricing
CLS431144
Corning
®
Erlenmeyer cell culture flasks,
250 mL Erlenmeyer Flask w/ Vent Cap, polycarbonate, sterile
Expand
View Pricing
CLS431145
Corning
®
Erlenmeyer cell culture flasks,
500 mL Erlenmeyer Flask w/ Vent Cap, polycarbonate, sterile, 25/cs
Expand
View Pricing
CLS431255
Corning
®
Erlenmeyer cell culture flasks,
2 L Erlenmeyer Flask w/ Vent Cap, polycarbonate, sterile, 6/cs
Expand
View Pricing
CLS431282
Corning
®
Erlenmeyer cell culture flasks,
capacity (3 L), PETG, sterile; γ-irradiated
Expand
View Pricing
CLS430183
Corning
®
Erlenmeyer cell culture flasks,
250 mL Erlenmeyer Flask w/ Flat Cap, polycarbonate, sterile, 50/cs
Expand
View Pricing
CLS430422
Corning
®
Erlenmeyer cell culture flasks,
500 mL Erlenmeyer Flask w/ Flat Cap, polycarbonate, sterile, 25/cs
Expand
View Pricing
CLS430421
Corning
®
Erlenmeyer cell culture flasks,
125 mL Erlenmeyer Flask w/ Flat Cap, polycarbonate, sterile, 50/cs
Expand
View Pricing
CLS431146
Corning
®
Erlenmeyer cell culture flasks,
1 L Erlenmeyer Flask w/ Flat Cap, polycarbonate, sterile, 25/cs
Expand
View Pricing
CLS431284
Corning
®
Erlenmeyer cell culture flasks,
capacity (5 L), PETG, sterile
Expand
View Pricing
CLS431280
Corning
®
Erlenmeyer cell culture flasks,
capacity (2 L), PETG, sterile; γ-irradiated
Expand
View Pricing