Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
cryogenic vials, internal thread
Products
Product Category
vials (10)
Color
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS430488
Corning
®
cryogenic vials, internal thread,
capacity 2.0 mL, bottom, round, seal, washer, self-standing, case of 500
Expand
View Pricing
CLS431386
Corning
®
cryogenic vials, internal thread,
capacity 2.0 mL, bottom, round, seal, washer, self-standing, case of 250
Expand
View Pricing
CLS430487
Corning
®
cryogenic vials, internal thread,
bottom, conical, capacity 1.2 mL, seal, washer, self-standing
Expand
View Pricing
CLS430489
Corning
®
cryogenic vials, internal thread,
capacity 2.0 mL, bottom, round, seal, washer, self-standing: no
Expand
View Pricing
CLS430656
Corning
®
cryogenic vials, internal thread,
bottom, round, capacity 5.0 mL, seal, washer, self-standing
Expand
View Pricing
CLS430491
Corning
®
cryogenic vials, internal thread,
capacity 4.0 mL, bottom, round, seal, washer, self-standing
Expand
View Pricing
CLS8672
Corning
®
cryogenic vials, internal thread,
capacity 2.0 mL, 1D barcodes
Expand
View Pricing
CLS430492
Corning
®
cryogenic vials, internal thread,
capacity 5.0 mL, bottom, round, seal, washer, self-standing: no
Expand
View Pricing
CLS430490
Corning
®
cryogenic vials, internal thread,
capacity 4.0 mL, bottom, round, seal, washer, self-standing: no
Expand
View Pricing
CLS8670
Corning
®
cryogenic vials, internal thread,
capacity 2.0 mL, 1D and 2D barcodes
Expand
View Pricing