Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed
Products
Product Category
pipettes (6)
Color
Manufacturer
Capacity (mL)
Length (mm)
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS4250
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed,
capacity 25 mL
Expand
View Pricing
CLS4100
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed,
capacity 10 mL
Expand
View Pricing
CLS4050
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed,
capacity 5 mL
Expand
View Pricing
CLS4500
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed,
capacity 50 mL
Expand
View Pricing
CLS4020
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed,
capacity 2 mL
Expand
View Pricing
CLS4010
Corning
®
Costar
®
Stripette
®
serological pipettes, bulk packed,
capacity 1 mL
Expand
View Pricing