Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
CoolCell
™
LX Cell Freezing Container
Products
Color
Manufacturer
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS432001
Corning
®
CoolCell
™
LX Cell Freezing Container,
(12 x 1 mL or 2 mL cryogenic vials), purple
Expand
View Pricing
CLS432002
Corning
®
CoolCell
™
LX Cell Freezing Container,
(12 x 1 mL or 2 mL cryogenic vials), green
Expand
View Pricing
CLS432004
Corning
®
CoolCell
™
LX Cell Freezing Container,
(12 x 1 mL or 2 mL cryogenic vials), pink
Expand
View Pricing
CLS432003
Corning
®
CoolCell
™
LX Cell Freezing Container,
(12 x 1 mL or 2 mL cryogenic vials), orange
Expand
View Pricing