Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Axygen
®
Deep Well Microplate
Products
Product Category
plates (5)
Manufacturer
Binding Type
Wells
Color
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
AXYP384240SQCS
Corning
®
Axygen
®
Deep Well Microplate,
size 384 wells, Not Treated, working volume 240 μL, pkg of 10x5plates/cs, sterile
Expand
View Pricing
AXYP384240SQC
Corning
®
Axygen
®
Deep Well Microplate,
size 384 wells, Not Treated, working volume 240 μL, pkg of 10x5plates/cs
Expand
View Pricing
AXYP384240SQCSI
Corning
®
Axygen
®
Deep Well Microplate,
indiv. wrapped, size 384 wells, working volume 240 μL, pkg of 50 ea, sterile
Expand
View Pricing
AXYP384120SQC
Corning
®
Axygen
®
Deep Well Microplate,
size 384 wells, working volume 120 μL, Not Treated, pkg of 10x5plates/cs
Expand
View Pricing
AXYP384120SQCS
Corning
®
Axygen
®
Deep Well Microplate,
size 384 wells, working volume 120 μL, Not Treated, pkg of 10x5plates/cs, sterile
Expand
View Pricing