Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 Well Storage Microplate
Products
Product Category
plates (9)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
CLS3365
Corning
®
96 Well Storage Microplate,
round bottom clear, polypropylene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3363
Corning
®
96 Well Storage Microplate,
V-bottom clear, polypropylene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3359
Corning
®
96 Well Storage Microplate,
round bottom clear, polypropylene, bag of 25 ×, sterile, lid: no
Expand
View Pricing
CLS3357
Corning
®
96 Well Storage Microplate,
V-bottom clear, polypropylene, bag of 25 ×, sterile, lid: no
Expand
View Pricing
CLS3356
Corning
®
96 Well Storage Microplate,
round bottom, black polypropylene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3344
Corning
®
96 Well Storage Microplate,
V-bottom, expanded volume, polypropylene, standard height, bag of 10 ×, sterile, lid: no
Expand
View Pricing
CLS3343
Corning
®
96 Well Storage Microplate,
V-bottom, expanded volume, polypropylene, standard height, bag of 10 ×, non-sterile, lid: no
Expand
View Pricing
CLS3364
Corning
®
96 Well Storage Microplate,
flat bottom clear, polypropylene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3355
Corning
®
96 Well Storage Microplate,
round bottom, white polypropylene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing