Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 Well Solid Polystyrene Microplate
Products
Product Category
plates (6)
Color
Manufacturer
Binding Type
Surface Area cm²
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS3917
Corning
®
96 Well Solid Polystyrene Microplate,
flat bottom, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, lid
Expand
View Pricing
CLS3915
Corning
®
96 Well Solid Polystyrene Microplate,
flat bottom, black polystyrene, plate of 100 (in bags of 25), non-sterile, lid: no
Expand
View Pricing
CLS3912
Corning
®
96 Well Solid Polystyrene Microplate,
flat bottom, white polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3916
Corning
®
96 Well Solid Polystyrene Microplate,
flat bottom, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing
CLS3362
Corning
®
96 Well Solid Polystyrene Microplate,
flat bottom, white polystyrene, bag of 25, sterile, lid: no
Expand
View Pricing
CLS3792
Corning
®
96 Well Solid Polystyrene Microplate,
round bottom, black polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing