Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 well Polypropylene Deep Well Plate
Products
Product Category
plates (6)
Manufacturer
Binding Type
Volume (ml)
Wells
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS3960
Corning
®
96 well Polypropylene Deep Well Plate,
V-bottom clear, polypropylene, bag of 5 ×, sterile, lid: no
Expand
View Pricing
CLS3958
Corning
®
96 well Polypropylene Deep Well Plate,
round bottom clear, polypropylene, bag of 5 ×, sterile, lid: no
Expand
View Pricing
CLS3961
Corning
®
96 well Polypropylene Deep Well Plate,
V-bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no
Expand
View Pricing
CLS3956
Corning
®
96 well Polypropylene Deep Well Plate,
V-bottom clear, polypropylene, bag of 10 ×, sterile, lid: no
Expand
View Pricing
CLS3959
Corning
®
96 well Polypropylene Deep Well Plate,
round bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no
Expand
View Pricing
CLS3957
Corning
®
96 well Polypropylene Deep Well Plate,
V-bottom clear, polypropylene, bag of 10 ×, non-sterile, lid: no
Expand
View Pricing