Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper(II) phthalocyanine
Synonym(s):
CuPc, Phthalocyanine blue, Pigment Blue 15
Empirical Formula (Hill Notation):
C
32
H
16
CuN
8
CAS Number:
147-14-8
Molecular Weight:
576.07
EC Number:
205-685-1
Products
Brand
Sigma-Aldrich (5)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Markush Class
Markush Group
Special Grade
Sublimed grade (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
546682
Copper(II) phthalocyanine,
Dye content >99 %
Expand
View Pricing
546674
Copper(II) phthalocyanine,
sublimed grade, Dye content 99 %
Expand
View Pricing
252980
Copper(II) phthalocyanine,
β
-form, Dye content 90 %
Expand
View Pricing
932515
Copper(II) phthalocyanine,
≥98%
Expand
View Pricing
702854
Copper(II) phthalocyanine,
triple-sublimed grade, >99.95% trace metals basis
Expand
View Pricing