Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Conjugated Polymer Nanoparticle (CPN)
Products
Product Category
kits (192)
Brand
Sigma-Aldrich (192)
Package Size
Filter Products
Showing 1-20 of 192
Page 1 of 10
Page 1 of 10
Compare
Product Number
Description
Pricing
5300NG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
530 nm (green), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
1000MG5
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
1000 nm (IR-II), maleimide-thiol functionalized, 1 ea of 10 μg antibody kit
Expand
View Pricing
4750MG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
475 nm (blue), maleimide-thiol functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
6800NG7
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
680 nm (red), carboxy-NHS-amine functionalized, 1 ea of 100 μg antibody kit
Expand
View Pricing
5300MG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
530 nm (green), maleimide-thiol functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
4200NG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
420 nm (violet), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
6600MG5
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
660 nm (red), maleimide-thiol functionalized, 1 ea of 10 μg antibody kit
Expand
View Pricing
1000NG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
1000 nm (IR-II), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
8400NG7
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
840 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 100 μg antibody kit
Expand
View Pricing
4200MN7
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
420 nm (violet), maleimide-thiol functionalized, 1 ea of 60 nmol oligonucleotide kit
Expand
View Pricing
6800MG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
680 nm (red), maleimide-thiol functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
6100NG5
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
610 nm (orange), carboxy-NHS-amine functionalized, 1 ea of 10 μg antibody kit
Expand
View Pricing
4350NG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
435 nm (indigo), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
1000MN6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
1000 nm (IR-II), maleimide-thiol functionalized, 1 ea of 18 nmol oligonucleotide kit
Expand
View Pricing
8400NG5
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
840 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 10 μg antibody kit
Expand
View Pricing
4750NG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
475 nm (blue), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
5100NG6
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
510 nm (green), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Expand
View Pricing
510BMN5
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
510B nm (green), maleimide-thiol functionalized, 1 ea of 6 nmol oligonucleotide kit
Expand
View Pricing
6800MN5
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
680 nm (red), maleimide-thiol functionalized, 1 ea of 6 nmol oligonucleotide kit
Expand
View Pricing
4350MN7
Conjugated Polymer Nanoparticle (CPN),
fluorescence λ
em
435 nm (indigo), maleimide-thiol functionalized, 1 ea of 60 nmol oligonucleotide kit
Expand
View Pricing
Page 1 of 10
Page 1 of 10