Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Commercial bronze standard for optical emission and x-ray spectroscopic analysis
Products
Manufacturer
Format
Quality Segment
Special Grade
certified reference material (5)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
NIST1116
Commercial bronze standard for optical emission and x-ray spectroscopic analysis,
NIST
®
SRM
®
1116
Expand
View Pricing
NIST1117
Commercial bronze standard for optical emission and x-ray spectroscopic analysis,
NIST
®
SRM
®
1117
Expand
View Pricing
NISTC1115
Commercial bronze standard for optical emission and x-ray spectroscopic analysis,
NIST
®
SRM
®
C1115, A (block)
Expand
View Pricing
NISTC1117
Commercial bronze standard for optical emission and x-ray spectroscopic analysis,
NIST
®
SRM
®
C1117, C (block)
Expand
View Pricing
NIST1115
Commercial bronze standard for optical emission and x-ray spectroscopic analysis,
NIST
®
SRM
®
1115
Expand
View Pricing