Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Color Reference Solutions
Products
Brand
Supelco (12)
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
analytical standard (12)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
90232
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors B, BY, Y, GY, R
Expand
View Pricing
83952
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors B, BY, Y, GY, R
Expand
View Pricing
86293
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors BY
Expand
View Pricing
83883
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors Y
Expand
View Pricing
92936
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors B
Expand
View Pricing
72666
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors BY
Expand
View Pricing
83967
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors Y
Expand
View Pricing
83951
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors B
Expand
View Pricing
95872
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors R
Expand
View Pricing
87448
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors R
Expand
View Pricing
82995
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors GY
Expand
View Pricing
90269
Color Reference Solutions,
related to Ph.Eur Color intensity testing, analytical standard, Set of colors GY
Expand
View Pricing