
Cobalt iron oxide

Synonym(s):
Cobalt diiron tetraoxide, Cobalt ferrate, Cobalt ferrite, Cobalt iron spinel
Empirical Formula (Hill Notation):
CoFe2O4
CAS Number:
Molecular Weight:
234.62
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2