Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt(II) chloride
Synonym(s):
Cobalt dichloride, Cobalt muriate, Cobaltous chloride, Dichlorocobalt
Linear Formula:
CoCl
2
CAS Number:
7646-79-9
Molecular Weight:
129.84
EC Number:
231-589-4
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (2)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
232696
Cobalt(II) chloride,
97%
Expand
View Pricing
60818
Cobalt(II) chloride,
purum p.a., anhydrous, ≥98.0% (KT)
Expand
View Pricing
449776
Cobalt(II) chloride,
AnhydroBeads
™
, −10 mesh, 99.9% trace metals basis
Expand
View Pricing
409332
Cobalt(II) chloride,
AnhydroBeads
™
, −10 mesh, 99.995% trace metals basis
Expand
View Pricing