Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt/Chromium/Tungsten/Nickel/Iron/Manganese
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF09271863
Cobalt/Chromium/Tungsten/Nickel/Iron/Manganese,
rod, Co 50%/Cr 20%/W 15%/Ni 10%/Fe 3%/Mn 2%, 9.8 mm diameter, length 50 mm
Expand
View Pricing
GF71483924
Cobalt/Chromium/Tungsten/Nickel/Iron/Manganese,
rod, 9.8 mm diameter, Co50%/Cr20%/W 15%/Ni10%/Fe 3%/Mn 2%, length 200 mm
Expand
View Pricing
GF76797747
Cobalt/Chromium/Tungsten/Nickel/Iron/Manganese,
rod, 9.8 mm diameter, Co50%/Cr20%/W 15%/Ni10%/Fe 3%/Mn 2%, length 100 mm
Expand
View Pricing
GF81987711
Cobalt/Chromium/Tungsten/Nickel/Iron/Manganese,
rod, Co50%/Cr20%/W 15%/Ni10%/Fe 3%/Mn 2%, 9.8 mm diameter, length 500 mm
Expand
View Pricing