Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Clavies
®
autoclave bags
Products
Product Category
bags (9)
Manufacturer
Color
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
BAH131851224
Clavies
®
autoclave bags,
width × H × thickness 12 in. (300 mm) × 24 in. (610 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131852436
Clavies
®
autoclave bags,
width × thickness 24 in. (610 mm) × 36 in. (910 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131820812
Clavies
®
autoclave bags,
clear, pack of 100 ea
Expand
View Pricing
BAH131820810
Clavies
®
autoclave bags,
W × H × thickness 8 in. (200 mm) × 10 in. (250 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131820815
Clavies
®
autoclave bags,
clear, pack of 100 ea
Expand
View Pricing
BAH131821012
Clavies
®
autoclave bags,
width × H × thickness 10 in. (250 mm) × 12 in. (300 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131821015
Clavies
®
autoclave bags,
width × H × thickness 10 in. (250 mm) × 15 in. (380 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131852430
Clavies
®
autoclave bags,
width × H × thickness 24 in. (610 mm) × 30 in. (760 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131821010
Clavies
®
autoclave bags,
width × H × thickness 10 in. (250 mm) × 10 in. (250 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Expand
View Pricing