Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Citric acid trisodium salt
Linear Formula:
C
6
H
5
O
7
Na
3
CAS Number:
68-04-2
Molecular Weight:
258.07
EC Number:
200-675-3
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (1)
Feature
Formula Weight
Physical Form
Quality Segment
Special Grade
Anhydrous (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C3674
Citric acid trisodium salt,
anhydrous, ≥98% (GC)
Expand
View Pricing