Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chorionic gonadotropin human
Synonym(s):
Choriogonin, HCG
CAS Number:
9002-61-3
EC Number:
232-660-2
Products
Product Category
hormones (4)
peptides (4)
Brand
Sigma-Aldrich (4)
Biological Source
Gene Alias
Physical Form
Tissue
Quality Segment
Package Size
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CG10
Chorionic gonadotropin human,
lyophilized powder, vial of ~10,000 IU
Expand
View Pricing
C1063
Chorionic gonadotropin human,
lyophilized powder, vial of ~2,500 IU
Expand
View Pricing
CG5
Chorionic gonadotropin human,
lyophilized powder, vial of ~5,000 IU
Expand
View Pricing
C8554
Chorionic gonadotropin human,
lyophilized powder, suitable for mouse embryo cell culture
Expand
View Pricing