Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Choline hydroxide solution
Linear Formula:
HOCH
2
CH
2
N(CH
3
)
3
OH
CAS Number:
123-41-1
Molecular Weight:
121.18
Products
Product Category
organic bases (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
292257
Choline hydroxide solution,
46 wt. % in H
2
O
Expand
View Pricing
333018
Choline hydroxide solution,
45 wt. % in methanol
Expand
View Pricing