Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chitin from shrimp shells
Synonym(s):
Poly-(1→4)-β-N-acetyl-D-glucosamine
CAS Number:
1398-61-4
EC Number:
215-744-3
Products
Product Category
carbohydrates (2)
polysaccharides (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Physical Form
Product Line
Quality Segment
Optical Isomer
Special Grade
Practical (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
C7170
Chitin from shrimp shells,
practical grade, powder
Expand
View Pricing
C9752
Chitin from shrimp shells,
BioReagent, suitable for analysis of chitinase, purified powder
Expand
View Pricing