
Certified Reference Material Fatty Acid Methyl Ester (FAME) Standard

Synonym(s):
Diesel
CAS Number:
EC Number:
Products
Product Category
Manufacturer
Physical Form
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing