Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CERTAN
®
capillary bottle
Synonym(s):
Capillary bottle CERTAN
®
Products
Product Category
bottles (3)
vials (3)
Brand
Supelco (2)
Diameter (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
44419
CERTAN
®
capillary bottle,
1.5 mL bottle, diam. × H 12 mm × 50 mm
Expand
View Pricing
44420
CERTAN
®
capillary bottle,
4.5 ml bottle, diam. × H 16 mm × 71 mm
Expand
View Pricing
44421
CERTAN
®
capillary bottle,
10 ml bottle, diam. × H 30 mm × 71 mm
Expand
View Pricing