Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cd-based Core/Shell Quantum Dots with Streptavidin
Products
Product Category
quantum dots (10)
Brand
Sigma-Aldrich (10)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
OCNQSS525
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
525 nm, 1 μM
Expand
View Pricing
OCNQSS450
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
450 nm, 1 μM
Expand
View Pricing
OCNQSS665
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
665 nm, 1 μM
Expand
View Pricing
OCNQSS620
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
620 nm, 1 μM
Expand
View Pricing
OCNQSS645
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
645 nm, 1 μM
Expand
View Pricing
OCNQSS490
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
490 nm, 1 μM
Expand
View Pricing
OCNQSS580
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
580 nm, 1 μM
Expand
View Pricing
OCNQSS560
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
560 nm, 1 μM
Expand
View Pricing
OCNQSS600
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
600 nm, 1 μM
Expand
View Pricing
OCNQSS540
Cd-based Core/Shell Quantum Dots with Streptavidin,
fluorescence λ
em
540 nm, 1 μM
Expand
View Pricing