Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cationic Liposomes for DNA/RNA delivery
Products
Brand
Sigma-Aldrich (39)
Chemical Composition
Quality Segment
Filter Products
Showing 1-20 of 39
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GEN-7000
Cationic Liposomes for DNA/RNA delivery,
DOTAP (100%)
Expand
View Pricing
GEN-7019
Cationic Liposomes for DNA/RNA delivery,
DOTMA:DOPE (1:1)
Expand
View Pricing
GEN-7002
Cationic Liposomes for DNA/RNA delivery,
DOTAP:DOPE (1:1)
Expand
View Pricing
GEN-7001
Cationic Liposomes for DNA/RNA delivery,
DOTAP:Cholesterol (1:1)
Expand
View Pricing
GEN-7009
Cationic Liposomes for DNA/RNA delivery,
DDAB:Cholesterol (1:1)
Expand
View Pricing
GEN-7007
Cationic Liposomes for DNA/RNA delivery,
DOTAP (100%) containing 0.5% DiO (Fluorescent)
Expand
View Pricing
GEN-7020
Cationic Liposomes for DNA/RNA delivery,
DOTMA (100%)
Expand
View Pricing
GEN-7006
Cationic Liposomes for DNA/RNA delivery,
DOTAP:DOPE (1:1) containing 0.5% Rhod-PE (Fluorescent)
Expand
View Pricing
GEN-7015
Cationic Liposomes for DNA/RNA delivery,
DDAB:DOPE (1:1) containing 0.5% Rhod-PE (Fluorescent)
Expand
View Pricing
GEN-7036
Cationic Liposomes for DNA/RNA delivery,
DOTAP:Chol:DOPE (1:0.75:0.5)
Expand
View Pricing
GEN-7024
Cationic Liposomes for DNA/RNA delivery,
DOTMA:DOPE (1:1) containing 0.5% Rhod-PE (Fluorescent)
Expand
View Pricing
GEN-7029
Cationic Liposomes for DNA/RNA delivery,
DC-Chol:DOPE (1:1) containing 0.5% NBD-DOPE (Fluorescent)
Expand
View Pricing
GEN-7028
Cationic Liposomes for DNA/RNA delivery,
DC-Chol:DOPE (1:2)
Expand
View Pricing
GEN-7003
Cationic Liposomes for DNA/RNA delivery,
DOTAP:Cholesterol (1:1) containing 0.5% NBD-DOPE (Fluorescent)
Expand
View Pricing
GEN-7017
Cationic Liposomes for DNA/RNA delivery,
DDAB (100%) containing 0.5% Rhod-PE (Fluorescent)
Expand
View Pricing
GEN-7018
Cationic Liposomes for DNA/RNA delivery,
DOTMA:Cholesterol (1:1)
Expand
View Pricing
GEN-7030
Cationic Liposomes for DNA/RNA delivery,
DC-Chol:DOPE (1:1) containing 0.5% Rhod-PE (Fluorescent)
Expand
View Pricing
GEN-7032
Cationic Liposomes for DNA/RNA delivery,
DC-Chol:DOPE (1:2) containing 0.5% Rhod-PE (Fluorescent)
Expand
View Pricing
GEN-7008
Cationic Liposomes for DNA/RNA delivery,
DOTAP (100%) containing 0.5% DiI (Fluorescent)
Expand
View Pricing
GEN-7005
Cationic Liposomes for DNA/RNA delivery,
DOTAP:DOPE (1:1) containing 0.5% NBD-DOPE (Fluorescent)
Expand
View Pricing
Page 1 of 2
Page 1 of 2