Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Catalase from bovine liver
Synonym(s):
H
2
O
2
:H
2
O
2
oxidoreductase, Catalase from bovine liver
CAS Number:
9001-05-2
EC Number:
232-577-1
Products
Product Category
enzymes (7)
peptides (1)
Brand
Sigma-Aldrich (8)
Biological Source
pH Value
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
C9322
Catalase from bovine liver,
lyophilized powder, 2,000-5,000 units/mg protein
Expand
View Pricing
C40
Catalase from bovine liver,
lyophilized powder, ≥10,000 units/mg protein
Expand
View Pricing
C1345
Catalase from bovine liver,
powder, suitable for cell culture, 2,000-5,000 units/mg protein
Expand
View Pricing
C30
Catalase from bovine liver,
aqueous suspension, 10,000-40,000 units/mg protein
Expand
View Pricing
C100
Catalase from bovine liver,
aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Expand
View Pricing
C3155
Catalase from bovine liver,
aqueous solution, ≥30,000 units/mg protein
Expand
View Pricing
SRE0041
Catalase from bovine liver,
Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, 2,000-5,000 units/mg protein
Expand
View Pricing
E3289
Catalase from bovine liver,
≥20000 units/mg protein, lyophilized powder
Expand
View Pricing