Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carboxen
®
-1010 PLOT Capillary GC Column
EC Number:
262-373-8
Products
Product Category
columns (2)
Brand
Supelco (2)
Agency
Column Type
ID (mm)
Length (m)
Application
d
f
(µm)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
25467
Carboxen
®
-1010 PLOT Capillary GC Column,
L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Expand
View Pricing
24246
Carboxen
®
-1010 PLOT Capillary GC Column,
L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Expand
View Pricing