Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbonic Anhydrase from bovine erythrocytes
Synonym(s):
Carbonate Dehydratase, Carbonate Hydrolyase, Carbonic Anhydrase from bovine erythrocytes
CAS Number:
9001-03-0
EC Number:
232-576-6
Products
Product Category
enzymes (4)
Brand
Sigma-Aldrich (5)
Biological Source
Physical Form
Product Line
Purity
Tissue
Quality Segment
Application
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C3934
Carbonic Anhydrase from bovine erythrocytes,
lyophilized powder, ≥2,000 W-A units/mg protein
Expand
View Pricing
C2624
Carbonic Anhydrase from bovine erythrocytes,
≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Expand
View Pricing
C7025
Carbonic Anhydrase from bovine erythrocytes,
BioReagent, suitable for GFC marker
Expand
View Pricing
C2273
Carbonic Anhydrase from bovine erythrocytes,
For use as a marker in SDS-PAGE
Expand
View Pricing
C5024
Carbonic Anhydrase from bovine erythrocytes,
non-denaturing PAGE marker
Expand
View Pricing