Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-tri-Calcium phosphate
Synonym(s):
β-TCP, β-Tricalcium phosphate
Empirical Formula (Hill Notation):
Ca
3
O
8
P
2
CAS Number:
7758-87-4
Molecular Weight:
310.18
EC Number:
231-840-8
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Special Grade
Puriss (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
49963
β-tri-Calcium phosphate,
puriss. p.a., ≥98% β-phase basis (sintered Powder)
Expand
View Pricing
13204
β-tri-Calcium phosphate,
puriss. p.a., ≥98% β-phase basis (unsintered powder)
Expand
View Pricing