Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brilliant Blue R
Synonym(s):
Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R
Empirical Formula (Hill Notation):
C
45
H
44
N
3
NaO
7
S
2
CAS Number:
6104-59-2
Molecular Weight:
825.97
EC Number:
228-060-5
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
27816
Brilliant Blue R,
250, for microscopy
Expand
View Pricing
B7920
Brilliant Blue R,
≥70% purity (HPLC), powder
Expand
View Pricing
B0149
Brilliant Blue R,
Dye content ~50 %, Powder
Expand
View Pricing