Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Titrette
®
digital bottle-top burette
Products
Product Category
burettes (6)
Manufacturer
Capacity (mL)
Volume (ml)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR4760161
BRAND
®
Titrette
®
digital bottle-top burette,
capacity 50 mL, without interface RS 232
Expand
View Pricing
BR4760151
BRAND
®
Titrette
®
digital bottle-top burette,
capacity 25 mL, without interface RS 232
Expand
View Pricing
Z567132
BRAND
®
Titrette
®
digital bottle-top burette,
volume 50 mL, accuracy: 0.06%, with interface RS 232
Expand
View Pricing
BR4760141
BRAND
®
Titrette
®
digital bottle-top burette,
capacity 10 mL, without interface RS 232
Expand
View Pricing
Z567124
BRAND
®
Titrette
®
digital bottle-top burette,
volume 25 mL, accuracy: 0.07%, with interface RS 232
Expand
View Pricing
BR4760241
BRAND
®
Titrette
®
digital bottle-top burette,
capacity 10 mL, with interface RS 232
Expand
View Pricing