Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
stopper
Products
Product Category
stoppers (12)
Manufacturer
Diameter (mm)
Height (mm)
Color
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
BR144382
BRAND
®
stopper,
red rubber, H 20 mm, top diam. 12 mm
Expand
View Pricing
BR144388
BRAND
®
stopper,
red rubber, H 25 mm, top diam. 22 mm
Expand
View Pricing
BR144315
BRAND
®
stopper,
silicone, H 20 mm, top diam. 14 mm
Expand
View Pricing
BR144385
BRAND
®
stopper,
red rubber, H 20 mm, top diam. 16 mm
Expand
View Pricing
BR144310
BRAND
®
stopper,
silicone, H 20 mm, top diam. 12 mm
Expand
View Pricing
BR144386
BRAND
®
stopper,
red rubber, H 20 mm, top diam. 18 mm
Expand
View Pricing
BR144325
BRAND
®
stopper,
silicone, H 20 mm, top diam. 18 mm
Expand
View Pricing
BR144340
BRAND
®
stopper,
silicone, H 30 mm, top diam. 27 mm
Expand
View Pricing
BR144320
BRAND
®
stopper,
silicone, H 20 mm, top diam. 16 mm
Expand
View Pricing
BR144384
BRAND
®
stopper,
red rubber, H 20 mm, top diam. 14 mm
Expand
View Pricing
BR144330
BRAND
®
stopper,
silicone, H 25 mm, top diam. 22 mm
Expand
View Pricing
BR144390
BRAND
®
stopper,
red rubber, H 30 mm, top diam. 27 mm
Expand
View Pricing