Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
PCR tubes, strips of 8
Products
Product Category
tubes (6)
Manufacturer
Binding Type
Capacity (mL)
Working Volume (ml)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR781332
BRAND
®
PCR tubes, strips of 8,
attached single caps, standard profile, capacity 0.2 mL
Expand
View Pricing
BR781316
BRAND
®
PCR tubes, strips of 8,
attached transparent single caps, standard profile, capacity 0.2 mL, white wells
Expand
View Pricing
BR781333
BRAND
®
PCR tubes, strips of 8,
attached single caps, low profile, capacity 0.15 mL
Expand
View Pricing
BR781315
BRAND
®
PCR tubes, strips of 8,
Expand
View Pricing
BR781318
BRAND
®
PCR tubes, strips of 8,
Expand
View Pricing
BR781317
BRAND
®
PCR tubes, strips of 8,
attached transparent single caps, low profile, capacity 0.15 mL, white wells
Expand
View Pricing