Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
Products
Product Category
inserts (6)
Manufacturer
Binding Type
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR782826
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
,
smooth-walled, PC membrane, pore size 1 μm
Expand
View Pricing
BR782806
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
,
smooth-walled, PC membrane, pore size 0.4 μm
Expand
View Pricing
BR782846
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
,
smooth-walled, PC membrane, pore size 3 μm
Expand
View Pricing
BR782866
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
,
smooth-walled, PC membrane, pore size 8 μm
Expand
View Pricing
BR782816
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
,
smooth-walled, PET membrane, pore size 0.4 μm
Expand
View Pricing
BR782876
BRAND
®
Insert-Single 13mm for 24-/6-well BRANDplates
®
,
smooth-walled, PET membrane, pore size 8 μm
Expand
View Pricing