Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass
Products
Product Category
flasks (15)
Brand
Sigma-Aldrich (1)
Manufacturer
Capacity (mL)
Closure Type
Glassware Class
Package Size
Sterility
Style
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Z326852
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 10 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19
Expand
View Pricing
Z326917
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 100 mL, accuracy: 0.08 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
Z326895
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 50 mL, accuracy: 0.10 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
Z326887
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 50 mL, accuracy: 0.06 mL, neck joint: ST/NS 12/21, glass stopper
Expand
View Pricing
Z326860
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 20 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing
Z326933
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 250 mL, accuracy: 0.15 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
Z326941
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 500 mL, accuracy: 0.25 mL, neck joint: ST/NS 19/26, glass stopper
Expand
View Pricing
Z326844
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 5 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing
Z627429
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 25 mL, accuracy: 0.06 mL, neck joint: ST/NS 12/21, glass stopper
Expand
View Pricing
Z326968
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 1,000 mL, accuracy: 0.4 mL, neck joint: ST/NS 24/29, glass stopper
Expand
View Pricing
Z326909
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 100 mL, accuracy: 0.08 mL, neck joint: ST/NS 12/21, glass stopper
Expand
View Pricing
Z326925
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 200 mL, accuracy: 0.15 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
Z326984
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 5,000 mL, accuracy: 1.2 mL, neck joint: ST/NS 34/35
Expand
View Pricing
Z326976
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 2,000 mL, accuracy: 0.6 mL, neck joint: ST/NS 29/32, glass stopper
Expand
View Pricing
Z326879
BRAND
®
BLAUBRAND
®
volumetric flask, glass stopper, clear glass,
volume 25 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing