Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Nicotinamide adenine dinucleotide
Synonym(s):
β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, Nadide
Empirical Formula (Hill Notation):
C
21
H
27
N
7
O
14
P
2
CAS Number:
53-84-9
Molecular Weight:
663.43
EC Number:
200-184-4
Products
Product Category
nucleotides (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Gene Alias
Physical Form
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
N8285
β-Nicotinamide adenine dinucleotide,
pkg of 10 mg (per vial)
Expand
View Pricing
N8535
β-Nicotinamide adenine dinucleotide,
pkg of 50 mg (per vial)
Expand
View Pricing
N8410
β-Nicotinamide adenine dinucleotide,
pkg of 20 mg (per vial)
Expand
View Pricing